Learn More
Macherey-Nagel™ NucleoSpin™ Soil
Isolate DNA from soil. Macherey Nagel™ NucleoSpin™ enables soil Isolation of total DNA from diverse soil types
Brand: Macherey-Nagel™ 740780.50
229.33 GBP valid until 2024-12-31
Use promo code "23283" to get your promotional price.
This item is not returnable.
View return policy
Description
Features- Two alternative lysis buffers and a special additive (Enhancer SX) for optimal processing of various soil samples – high yield and high purity
- NucleoSpin™ Bead Tubes with ceramic beads for most efficient cell disruption of Gram-positive and Gram-negative bacteria, archaea, yeast, fungi, and algae in soil, sludge,or sediment
- NucleoSpin™ Inhibitor Removal Column to eliminate all PCR inhibitors, such as humic substances – DNA is ready-to-use for PCR without dilution
- Total DNA from microorganisms in soil and sediment
- Quantify and detect microorganisms from environmental samples
- Typical downstream applications: PCR, real-time PCR, Southern blotting, microarray technology
Specifications
Mini Spin Column | |
For Total DNA From Microorganisms in Soil and Sediment | |
90min. |
Silica Membrane Technology | |
50 Preps | |
2 to 10μg |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate; guanidine hydrochloride, 2-propanol