missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Soil

Isolate DNA from soil. Macherey Nagel™ NucleoSpin™ enables soil Isolation of total DNA from diverse soil types

Brand:  Macherey-Nagel™ 740780.10


 View more versions of this product

Product Code. 12979884

  • £119.00 / 10 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features
  • Two alternative lysis buffers and a special additive (Enhancer SX) for optimal processing of various soil samples – high yield and high purity
  • NucleoSpin™ Bead Tubes with ceramic beads for most efficient cell disruption of Gram-positive and Gram-negative bacteria, archaea, yeast, fungi, and algae in soil, sludge,or sediment
  • NucleoSpin™ Inhibitor Removal Column to eliminate all PCR inhibitors, such as humic substances – DNA is ready-to-use for PCR without dilution
Applications
  • Total DNA from microorganisms in soil and sediment
  • Quantify and detect microorganisms from environmental samples
  • Typical downstream applications: PCR, real-time PCR, Southern blotting, microarray technology
Specifications

Specifications

Mini Spin Column
For Total DNA From Microorganisms in Soil and Sediment
90min.
Silica Membrane Technology
10 Preps
2 to 10μg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate; guanidine hydrochloride, 2-propanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.