missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 1.8mL Externally-Threaded Universal Tubes and Racks
GREENER_CHOICE

Use the automation friendly tubes to improve the efficiency and throughput of your sample storage workflow. The latch rack offers ANSI SBS footprint compatible with automation platforms for liquid handling and sample storage. Tubes are available racked or in bulk, with 2D barcodes or non-coded. Empty racks are also available.

Brand:  Thermo Scientific™ 374502

640.03 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 13416113

  • £960.00 / Pack of 480

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Natural
Sterile SAL 10-6, Sterile
Natural
Flat Bottom
External Thread
White patch and graduations
Bulk
Externally-threaded Universal Tube
Nunc Universal tubes with 2D code
1.8 mL
Plastic
Cryobank
480/Cs.
2D on Tube Bottom
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1421966 Certificate TypeCertificate of Compliance Date24/01/2025 Catalog Number374502
Lot Number1420731 Certificate TypeCertificate of Compliance Date22/01/2025 Catalog Number374502
Lot Number1420700 Certificate TypeCertificate of Compliance Date20/01/2025 Catalog Number374502
Lot Number1420460 Certificate TypeCertificate of Compliance Date17/01/2025 Catalog Number374502
Lot Number1420290 Certificate TypeCertificate of Compliance Date12/01/2025 Catalog Number374502
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ 1.8mL Externally-Threaded Universal Tubes and Racks > Nunc 1.8mL External Thread Universal Tubes, w/ 2D code, non-racked bulk pack, 10 bags of 48 tubes, Sterile

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.