Learn More
Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Lower spending when storing compounds for small volume applications by using these 384-shallow well standard height polypropylene microplates.
£858.48 - £1375.00
Specifications
Barcode | No Barcode |
---|---|
Certifications/Compliance | Certified RNase and DNase Free. ANSI Compliant. |
Format | 384-well |
Lid | Without Lid |
Description | 384-well Polypropylene Sample Processing & Storage Plates |
Product Code | Brand | Color | Sterility | Volume (Metric) Well | Fit for Purpose | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Sterility | Volume (Metric) Well | Fit for Purpose | Price | Quantity & Availability | |||||
11341605
|
Thermo Scientific™
267459 |
Natural | Non-sterile | 55 μL | Certified Quality | N/A | ||||||
11351605
![]() |
Thermo Scientific™
267460 |
Natural | Sterile | 55 μL | Certified Quality |
£1320.00
Pack of 100 |
Estimated Shipment: 08-05-2025
Log in to see stock. |
|||||
10390135
![]() |
Thermo Scientific™
267461 |
Black | Non-sterile | 55 μL | Certified Quality |
£1375.00
Pack of 100 |
Estimated Shipment: 08-05-2025
Log in to see stock. |
|||||
10742024
![]() |
Thermo Scientific™
267462 |
White | Non-sterile | 55 μL | Certified Quality |
£1355.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors to allow for quick identification of storage plates.
Features:- Rounded wells are optimized for mixing and sample recovery
- Square rounded wells minimize wicking (capillary action)
- Natural plates for storage identification or storing colorimetric assays
- White plates for luminescent assays
- Black plates for fluorescent assays
- Customized bar coding is available
Specifications
No Barcode | |
384-well | |
384-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Nunc | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
55 μL | |
2 to 35 μL | |
Square, Round Bottom (U-well) | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
25 | |
Yes | |
ANSI/ SLAS Standard |
Certified RNase and DNase Free. ANSI Compliant. | |
Without Lid | |
16 x 24 | |
Low binding PP | |
384-well plate | |
Polypropylene | |
384 | |
Non-treated | |
Yes | |
100 | |
For Research Use Only. Not for use in diagnostic procedures. | |
Certified Quality |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.