missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 384 Well ShallowWell Standard Height Black and White Plates
Save costs in samples and reagents by reducing the total well volume to 25μL using Nunc 384-well shallow well standard height black and white polystyrene plates.
Brand: Thermo Scientific™ 264705
This item is not returnable.
View return policy
Description
Accommodate small sample volumes by using Thermo Scientific™ Nunc™ 384-Well ShallowWell Standard Height Black and White Plates with total well volume of 25μL, ideal for high throughput screenings. The standard plate height allows for easy adaptation to most automation systems.
Background, autofluorescence and crosstalk are improved when you use Nunc black or white microplates.- Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Without Lid | |
16 x 24 | |
Nunc | |
Microplate | |
Polystyrene | |
384 | |
Non-sterile | |
Flat Bottom (F-well) | |
25 |
Standard | |
384 Shallow Well Standard Height, black, non-treated, no lid, packaged 25/100 | |
100 Cases, 25 Pk. | |
Black | |
Low Flange Skirted | |
25 μL | |
2 to 20 μL | |
Non-treated | |
100 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.