missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 50 mL Bioreactor Tube

Product Code. 17790387

Product Code. 17790387

Brand: Thermo Scientific™ 332260

Only null left
Add to basket
Add to basket

Thermo Scientific™ Nunc™ Bioreactor Centrifuge Tubes are designed for high throughput protein expression screening and cell line development and optimization under suspension condition.

Thermo Scientific™ Nunc™ Bioreactor Centrifuge Tubes are designed for high throughput protein expression screening and cell line development and optimization under suspension condition. Adjustable 4-level vented cap with a 0.2 μm filter membrane allows for controlled gas exchange and air circulation without contamination. Ergonomic grooved cap for effortless handling and a large writing area for ease of identification.

  • Ideal for screening, small volume high throughput analysis
  • Protein expression suspension culture volumes ranging from 10 mL to 25 mL
  • Convenient pack sizes
  • Four different sizes of ventilation (dial type) on the cap.
  • Tube: Polypropylene
  • Cap: HDPE
  • Filter: PTFE
  • Dial: Polystyrene
  • 2 μm PTFE filter membrane
  • Suitable for suspension culture
  • Sterile - SAL 10-6
  • Non-pyrogenic
  • Non-cytotoxic
  • Shelf life: 2 years
TRUSTED_SUSTAINABILITY

Especificaciones

Closure Material HDPE
Capacity (Metric) Protein expression suspension culture volumes ranging from 10 mL to 25 mL
Disposable Single-use
Material Polypropylene
For Use With Ideal for screening, small volume high throughput analysis
Includes
  • Cap: HDPE
  • Filter: PTFE
  • Dial: Polystyrene 
  • 0.2 μm PTFE filter membrane
No. per Case 90
No. per Pack 3
Sterility Sterile
RNase Free Yes
Dnase Free Yes
Autoclavable Autoclavable
Bottom Shape Conical
Closure Type Quadruple-position Vent/Close Cap
Writing Area Yes
Product Type Conical Centrifuge Tube
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado