Learn More
Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates
Clone and replicate assays with these untreated polystyrene plates, suited for agglutination assays and sample collection.
Brand: Thermo Scientific™ 278606
Description
Store samples and perform agglutination assays with Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates, featuring transparent round bottom deep wells that hold up to 900μL per well.
- Alphanumeric grid provides quick and easy sample identification.
- Optimized for robotics and liquid handling systems
- Round-bottom wells for optimal sample recovery
- Untreated surface
- Sealable with well caps and sealing tape
- Customized bar coding is available
Specifications
Standard | |
8 x 12 | |
Nunc | |
Microplate | |
96 | |
Sterile | |
Round Bottom (U-well) | |
4 | |
128 x 86 x 41 mm |
Without Lid | |
32 Cases, 4 Pk. | |
Clear | |
Polystyrene | |
900 μL | |
Non-treated | |
32 | |
32/Cs. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.