missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates

Clone and replicate assays with these untreated polystyrene plates, suited for agglutination assays and sample collection.

Brand:  Thermo Scientific™ 278605

170.93 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10577131

  • £217.00 / Pack of 32
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Store samples and perform agglutination assays with Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates, featuring transparent round bottom deep wells that hold up to 900μL per well.

  • Alphanumeric grid provides quick and easy sample identification.
  • Optimized for robotics and liquid handling systems
  • Round-bottom wells for optimal sample recovery
  • Untreated surface
  • Sealable with well caps and sealing tape
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
Nunc
Microplate
96
Non-sterile
Round Bottom (U-well)
4
Without Lid
32 Cases, 4 Pk.
Clear
Polystyrene
900 μL
Non-treated
32
128 x 86 x 41 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number61104-05 Certificate TypeCertificate of Compliance Date06/12/2024 Catalog Number278605
Lot Number61104-02 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number278605
Lot Number63962 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number278605
Lot Number63967 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number278605
Lot Number62460 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number278605
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates > 96 DeepWell-1mL, Polystyrene, Non-Treated,without lid, Non-sterile

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.