missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates



Click to view available options
Color:
Black
Blue
Natural
Red
White
Yellow
Sterility:
Non-sterile
Sterile
Well Shape:
Round
Round, Conical Bottom (V-well)
Square, Conical Bottom (V-well)
Fit for Purpose:
Certified Quality
Sterile
Packungsgröße:
Pack of 120
Pack of 50
Pack of 60
Beschreibung
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
- Shared-wall design increases the well volume
- Wider wells and rounded bottom improve mixing
- Conical bottom well for optimal sample recovery
- Black and white plates available for fluorescent- and luminescent-based applications
- Low binding polypropylene for homogeneous assays and storage
Spezifikation
Spezifikation
| Barcode | No Barcode |
| Certifications/Compliance | Certified RNase and DNase Free. ANSI Compliant. |
| Format | Standard |
| Lid | Without Lid |
| Description | 96-well Polypropylene Sample Processing & Storage Plates |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Packaging | 5 pk., 60 cs. |
| Binding Type | Low binding PP |
| Product Line | Nunc |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur