missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
£143.23 - £630.55
Specifications
Format | 96-well |
---|---|
Lid | Without Lid |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Binding Type | Low binding PP |
Product Code | Brand | Color | Well Shape | Sterility | Fit for Purpose | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Well Shape | Sterility | Fit for Purpose | Price | Quantity & Availability | ||
10555321
|
Thermo Scientific™
249943 |
Red | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
£519.00
Pack of 120 |
In Stock
Log in to see stock. |
||
10525511
|
Thermo Scientific™
249944 |
Natural | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
£519.00
Pack of 120 |
In Stock
Log in to see stock. |
||
10634502
|
Thermo Scientific™
249945 |
Black | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
£361.00
Pack of 120 |
In Stock
Log in to see stock. |
||
10644502
|
Thermo Scientific™
249946 |
Natural | Round, Conical Bottom (V-well) | Sterile | Sterile |
£630.55
Pack of 120 |
In Stock
Log in to see stock. |
||
10602212
|
Thermo Scientific™
249947 |
Yellow | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
£251.00
Pack of 120 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||
10387491
|
Thermo Scientific™
249949 |
White | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
£351.00
Pack of 120 |
In Stock
Log in to see stock. |
||
10052401
|
Thermo Scientific™
249950 |
Blue | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
£364.00
Pack of 120 |
In Stock
Log in to see stock. |
||
10671722
|
Thermo Scientific™
267245 |
Natural | Round | Non-sterile | Certified Quality |
£476.78
Pack of 120 |
In Stock
Log in to see stock. |
||
10192161
|
Thermo Scientific™
267334 |
Natural | Round | Sterile | Certified Quality |
£614.04
Pack of 120 |
In Stock
Log in to see stock. |
||
10276031
|
Thermo Scientific™
267342 |
Black | Round | Non-sterile | Certified Quality |
£340.00
Pack of 120 |
In Stock
Log in to see stock. |
||
10796272
|
Thermo Scientific™
267350 |
White | Round | Non-sterile | Certified Quality |
£340.00
Pack of 120 |
In Stock
Log in to see stock. |
||
10234171
|
Thermo Scientific™
267385 |
Blue | Round | Non-sterile | Certified Quality |
£362.00
Pack of 120 |
In Stock
Log in to see stock. |
||
10355661
|
Thermo Scientific™
267407 |
Yellow | Round | Non-sterile | Certified Quality |
£235.00
Pack of 120 |
In Stock
Log in to see stock. |
||
10479092
|
Thermo Scientific™
442587 |
Natural | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
£294.12
Pack of 60 |
In Stock
Log in to see stock. |
||
10652672
|
Thermo Scientific™
267369 |
Red | Round | Non-sterile | Certified Quality |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11375850
|
Thermo Scientific™
95040410 |
Natural | Square, Conical Bottom (V-well) | Non-sterile | Certified Quality |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
- Shared wall design increases the well volume
- Wider wells and rounded bottom improve mixing
- Conical bottom well for optimal sample recovery
- Black and white plates available for fluorescent- and luminescent-based applications
- Low binding polypropylene for homogeneous assays and storage
Spécification
96-well | |
96-well Polypropylene Sample Processing & Storage Plates | |
Low binding PP | |
96-well plate | |
Polypropylene | |
Non-treated | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
Yes |
Without Lid | |
8 x 12 | |
Nunc | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
96 | |
Yes | |
For Research Use Only. Not for use in diagnostic procedures. | |
ANSI/ SLAS Standard |