Learn More
Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Perform automated applications more precisely with these round-bottom plates, which are available with and without cell culture treatment.
£126.00 - £595.00
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Product Line | Nunc |
Product Type | Microplate |
Material | Polystyrene |
Product Code | Brand | Surface Treatment | Lid | No. per Pack | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Lid | No. per Pack | Price | Quantity & Availability | |||
10398221
|
Thermo Scientific™
262162 |
Non-treated | Without Lid | 1 |
£126.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
15317036
|
Thermo Scientific™
262146 |
Non-treated | Without Lid | 135 |
£221.88
Pack of 135 |
In Stock
Log in to see stock. |
|||
10418522
|
Thermo Scientific™
268152 |
Non-treated | Without Lid | 10 |
£299.00
Pack of 180 |
In Stock
Log in to see stock. |
|||
10418623
|
Thermo Scientific™
268200 |
Non-treated | With Lid | 10 |
£595.00
Pack of 160 |
In Stock
Log in to see stock. |
|||
11371605
|
Thermo Scientific™
268200 |
Non-treated | With Lid | 10 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates. The clear and U shaped wells are ideal for solution-based assays (e.g. agglutination assays) and common serial dilutions, allowing clean and easy pipetting. Available in a variety of options for surfaces, sterility, closures, and packaging sizes.
- Clear polystyrene has excellent optical properties ideal for optical measurements
- Thermo Scientific™ Nunclon™ Delta is a hydrophilic surface treatment whereas the non-treated polystyrene is more hydrophobic
- Customized bar coding is available
Specifications
Standard | |
Nunc | |
Polystyrene | |
Non-treated |
8 x 12 | |
Microplate | |
96 | |
Round Bottom (U-well) |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.