Learn More
Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes
GREENER_CHOICEBIOPHARMA
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
£183.35 - £1955.00
Specifications
Material Polypropylene
RNase Free Yes
Product Line Nunc™
Dnase Free Yes
View More Specs
Products 36
Product Code Brand Bottom Shape Capacity (Metric) Thread Style Style Price Quantity & Availability  
Product Code Brand Bottom Shape Capacity (Metric) Thread Style Style Price Quantity & Availability  
10257752
GREENER_CHOICE
View Documents
Thermo Scientific™
374080
Conical (Self-standing) 0.5 mL Internal No Writing Surface, A-R Base
£1135.00
Pack of 960
10320013
GREENER_CHOICEBIOPHARMA
View Documents
Thermo Scientific™
374081
Conical (Self-standing) 1 mL Internal Writing Surface
£1640.00
Pack of 960
10544641
GREENER_CHOICEBIOPHARMA
View accessory products View Documents
Thermo Scientific™
375353
Conical (Self-standing) 1 mL External Writing Surface, A-R Base
£542.00
Pack of 500
11301665
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
375353
Conical (Self-standing) 1 mL External Writing Surface, A-R Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10030711
View Documents
Thermo Scientific™
375299
Round 1 mL External Writing Surface This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11391655
View Documents
Thermo Scientific™
375299
Round 1 mL External Writing Surface This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10545601
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
347597
Conical (Self-standing) 1 mL External No Writing Surface, A-R Base
£372.00
Pack of 500
11391625
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
347597
Conical (Self-standing) 1 mL External No Writing Surface, A-R Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10654721
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
377224
Conical (Self-standing) 1 mL Internal Writing Surface, A-R Base
£577.00
Pack of 500
11301675
View Documents
Thermo Scientific™
377224
Conical (Self-standing) 1 mL Internal Writing Surface, A-R Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10529881
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
366656
Conical (Self-standing) 1 mL Internal Writing Surface
£509.00
Pack of 500
11381645
View Documents
Thermo Scientific™
366656
Conical (Self-standing) 1 mL Internal Writing Surface This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10669071
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
375418
Round (Self-standing) 1.8 mL External Writing Surface, A-R Base
£437.00
Pack of 450
11311665
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
375418
Round (Self-standing) 1.8 mL External Writing Surface, A-R Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10534641
View accessory products View Documents
Thermo Scientific™
340711
Round 1.8 mL External Writing Surface This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11331625
View accessory products View Documents
Thermo Scientific™
340711
Round 1.8 mL External Writing Surface This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10080601
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
347627
Round (Self-standing) 1.8 mL External No Writing Surface, A-R Base
£241.00
Pack of 450
11301635
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
347627
Round (Self-standing) 1.8 mL External No Writing Surface, A-R Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10577391
GREENER_CHOICEBIOPHARMA
View accessory products View Documents
Thermo Scientific™
377267
Round (Self-standing) 1.8 mL Internal Writing Surface, A-R Base
£453.00
Pack of 450
11341675
GREENER_CHOICEBIOPHARMA
View accessory products View Documents
Thermo Scientific™
377267
Round (Self-standing) 1.8 mL Internal Writing Surface, A-R Base
£1955.00
Pack of 1800
10674511
BIOPHARMA
View accessory products View Documents Promotion Details
Thermo Scientific™
368632
Round (Self-standing) 1.8 mL Internal Writing Surface
£352.00
Pack of 450
10004220
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
363401
Round 1.8 mL Internal Writing Surface
£475.00
Pack of 500
11321645
View Documents
Thermo Scientific™
363401
Round 1.8 mL Internal Writing Surface This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10452802
BIOPHARMA
View Documents
Thermo Scientific™
351934
Round 1.8 mL Internal Writing Surface, No Cap
£934.00
Pack of 2500
10070731
View accessory products View Documents
Thermo Scientific™
379189
Round (Self-standing) 3.6 mL Internal Writing Surface, A-R Base
£256.00
Pack of 400
10789871
View accessory products View Documents
Thermo Scientific™
366524
Round 3.6 mL Internal Writing Surface
£587.00
Pack of 400
11361645
View Documents
Thermo Scientific™
366524
Round 3.6 mL Internal Writing Surface This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10173772
BIOPHARMA
View Documents
Thermo Scientific™
367997
Round 3.6 mL Internal Writing Surface, No Cap
£612.00
Pack of 1500
10633452
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
337516
Round (Self-standing) 4.5 mL External Writing Surface, A-R Base
£367.00
Pack of 300
11381615
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
337516
Round (Self-standing) 4.5 mL External Writing Surface, A-R Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10356331
GREENER_CHOICEBIOPHARMA
View accessory products View Documents
Thermo Scientific™
347643
Round (Self-standing) 4.5 mL External No Writing Surface, A-R Base
£234.00
Pack of 300
11311635
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
347643
Round (Self-standing) 4.5 mL External No Writing Surface, A-R Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10716922
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
379146
Conical (Self-standing) 4.5 mL Internal Writing Surface, A-R Base
£410.00
Pack of 300
11361675
View Documents
Thermo Scientific™
379146
Conical (Self-standing) 4.5 mL Internal Writing Surface, A-R Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11331645
View Documents
Thermo Scientific™
363452
Round 4.5 mL Internal Writing Surface This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10358131
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
363452
Round 4.5 mL Internal Writing Surface
£183.35
Pack of 300
Description
Description
Store samples from general cold storage to the vapor phase of liquid nitrogen with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, available with either internal or external threads.
Features:
  • Non-cytotoxic, non-mutagenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
Specifications
Specifications
Polypropylene
Yes
Nunc™
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes