Learn More
Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Solid Caps
BIOPHARMA
Achieve maximum adhesion for a broad range of cell types.
£137.00 - £244.00
Specifications
Material Polystyrene
Sterility Sterile
View More Specs
Products 4
Product Code Brand Culture Area Closure Type Price Quantity & Availability  
Product Code Brand Culture Area Closure Type Price Quantity & Availability  
10068780
BIOPHARMA
View Documents
Thermo Scientific™
153732
80 cm² Solid
£244.00
Pack of 50
In Stock
to see stock.
Edge
Add to basket
13245219
View Documents
Thermo Scientific™
156505
175 cm² Vent/Close
£137.00
Pack of 32
In Stock
to see stock.
Add to basket
10354451
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
156502
175 cm² Solid
£216.00
Pack of 32
In Stock
to see stock.
Edge
Add to basket
10490033
View accessory products View Documents
Thermo Scientific™
163371
25 cm² Solid This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Solid Caps. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth. The vent/close caps with Y marks allow easy visual verification of venting position while promoting sufficient gas exchange.
Features:
  • Nunclon Delta Certification including testing on four different cell types is provided in each case.
  • Short wide necks, straight side walls and 1/3-turn caps are optimized for robotic operation.
  • Vent/close caps with “Y” marks - any of the legs in the “Y” pointing up indicates cap in venting position.
  • Sterilized by gamma irradiation and certified to be non-pyrogenic.
Warranty: 90 days
Specifications
Specifications
Polystyrene
Sterile
Videos
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.