Learn More
Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks
Perform large cell mass applications such as drug screening and scale up with these cell-culture treated TripleFlasks.
Brand: Thermo Scientific™ 132913
301.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 82
Additional Details : Weight : 5.87700kg
Description
Perform cell culture expansion and experience 500cm2 total growth area within the footprint of a standard flask using Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. TripleFlask receives the Nunclon™ Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types.
Three layers of growth surfaces provide a total culture area of 500 cm2 in the same footprint as a standard 175cm2 flask.
- Recommended working volume is 200mL
- Promotes conventional, flat, monolayer culturing
- External dimensions of 175cm2
- Also offered barcoded with large Code 128
Includes:
Nunclon Delta Certification including testing on four different cell lines is included in each case.
Specifications
No | |
Straight | |
1 | |
Sterile | |
500 cm2 | |
200 mL |
Cell Culture Treated TripleFlasks | |
32 | |
Same external dimensions as the standard Nunc T175 with straight neck | |
Filter | |
Cell Culture-Treated |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.