Learn More
Thermo Scientific™ Nunc™ Cell Factory™ Systems
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems.
Brand:  Thermo Scientific™ 164327
356.25 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 82
Additional Details : Weight : 5.70000kg
 View more versions of this product
Product Code. 10049040
  • £402.00 / Pack of 2
In Stock
to see stock.
Add to basket
Description
Description
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.
  • Ideal for adherent cells
  • Boost production efficiency with more surface area in a small footprint
  • Reduce contamination risks with a closed system
Specifications
Specifications
Original Cell Factory
2
Cell Factory System
Flat
6320 cm2
10
Polystyrene
1
Nunclon™ Delta
Adapter Cap and Port Cover Cap
Nunclon Delta
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.