Learn More
Thermo Scientific™ Nunc™ Cell Factory™ Systems
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems.
Brand: Thermo Scientific™ 139446
Code : 82
Additional Details : Weight : 18.64000kg
Description
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.
- Ideal for adherent cells
- Boost production efficiency with more surface area in a small footprint
- Reduce contamination risks with a closed system
Specifications
Original Cell Factory | |
2 | |
Cell Factory System | |
Flat | |
25280 cm2 | |
40 |
Polystyrene | |
1 | |
Nunclon™ Delta | |
Adapter Cap and Port Cover Cap | |
Cell Culture-Treated |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.