Learn More
Thermo Scientific™ Nunc™ IVF Center Well Dish
Experience consistently reliable, high-quality results during embryo and oocyte washing with Thermo Scientific™ Nunc Center Well Dishes.
Brand: Thermo Scientific™ 150260
Description
Thermo Scientific™ Nunc Center Well Dishes provide consistently reliable, IVF-quality results during embryo and oocyte washing.
Innovative features for quality and ease-of-use:
- Perfect angle of reservoir well walls for locating and easy washing of samples
- True flat bottom for maintaining optimal temperature
- Easy grip design for increased safety
Rigorously tested, certified and validated for IVF:
- CE-marked (Notified Body 2797), FDA-listed
- Produced under ISO 13485 with full lot traceability
Certificates of conformity for your assurance on every lot:
- Sterile (SAL 10-6)
- Non-pyrogenic (LAL test)
- USP Class VI test
- Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA)
Warranty: 90 days
Specifications
CE-marked, FDA-cleared, MEA-tested | |
Flat | |
Polystyrene (USP Class VI) | |
Non-treated | |
1 | |
1 | |
55 mm |
Center Well Dish, Lid w/Airvent | |
Clear | |
Sterile | |
Dish | |
120 | |
16 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.