missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Cryobank and Bank-It™ Replacement Caps
GREENER_CHOICE

Ensure contamination-free cap replacement. Closures are supplied racked for manual or automated replacement.

Brand:  Thermo Scientific™ 374018

137.00 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10513724

  • £241.00 / Pack of 960
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ensure contamination-free cap replacement with Thermo Scientific™ Nunc™ Cryobank and Bank-it™ Caps, which are supplied racked for manual or automated replacement. Available in natural, green, red or blue for simplified sample tracking.

Warranty: 90 days

  • Racked caps for automated or manual replacement of used caps
  • Racked in 96 microplate pattern for easy contamination free pick
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green
Non-sterile
2D Barcode
Green (Cap)
96
0.5mL and 1.0mL Cryobank and Bank-it Tubes
Polypropylene
Closure
Cryobank, Bank-It
10 racks of 96/case
960
Nunc™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.