Learn More
Thermo Scientific™ Nunc™ Cryobank and Bank-It™ Freezer Rack
Hold nineteen 0.5 mL cryobank racks or thirteen 1.0 mL cryobank racks with the Thermo Scientific™ Nunc™ Cryobank and Bank-It™ Freezer Rack.
Brand:  Thermo Scientific™ 367014
367.88 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10522964
  • £547.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Hold nineteen 0.5mL cryobank racks or thirteen 1.0mL cryobank racks with the Thermo Scientific™ Nunc™ Cryobank and Bank-It™ Vial Rack.
This verticle aluminum freezer rack with 74 cm in height is specifically designed to hold Thermo Scientific Nunc Cryobank in lock racks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cryobank and Bank-It Vial Rack
19 x 0.5mL or 13 x 1.0mL cryobank racks; one each
Cryobank and Bank-It Vial Rack
Cryobank
19 x 0.5mL or 13 x 1.0mL cryobank racks
0.5 mL (13 Cryobank Rack), 1 mL (13 Cryobank Rack)
Aluminum
2D Barcode
Silver