Learn More
Thermo Scientific™ Nunc™ CryoTube™ Colored Caps
Color-code your sample vials with replacement polypropylene screw caps featuring a silicone gasket to provide the best seal for internally threaded cryotube vials.
Brand: Thermo Scientific™ 336094
429.99 GBP valid until 2024-12-31
Use promo code "21935" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.80000kg
Alert:
Do not use CryoTubes in the liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex™ Tubing (Cat. No. 343958). Improper use may cause liquefied nitrogen to be trapped inside the vial and lead to pressure build-up, resulting in possible explosion or biohazard release. Use appropriate safety procedures as outlined in the Nunc Cryopreservation Manual when handling and disposing of the CryoTubes.
Description
Color-code your sample vials. Thermo Scientific™ Nunc™ CryoTube™ Colored Caps are for use with Nunc internally threaded CryoTube vials and include a silicone gasket to provide the best seal. Nunc CryoTubes are the most widely used cryo vials in the world, stemming from the first cryo vials manufactured in the early 1960s. The well-proven design, the high certification level and the large portfolio of vials provide researchers with a choice of the vial best suited for their application and peace of mind that the sample can be retrieved viable and unharmed.
- These replacement polypropylene screw caps have a silicone gasket to provide the best seal
- Available nonsterile or irradiated
Specifications
Assorted | |
Sterile | |
250/Pk., 2500/Cs. |
Polypropylene | |
Nunc | |
Croygenic Tube Closure |