Learn More
Thermo Scientific™ Nunc™ Deep Form Bacteriology Petri Dishes
GREENER_CHOICE
Culture fungi, bacteria and other microorganisms with these petri dishes
£10.15 - £421.00
Specifications
Sterility Sterile
View More Specs
Products 4
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
15300484
GREENER_CHOICE
View Documents
Thermo Scientific™
4014-12
150 mm
£312.00
Pack of 72
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
10765954
View Documents
Thermo Scientific™
4031
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15367036
View Documents
Thermo Scientific™
4031
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10581495
View Documents
Thermo Scientific™
4036
60 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc Lab-Tek Disposable Petri Dishes.
Thermo Scientific Nunc Cell Culture products offer solutions for cell growth and analysis and have been used by researchers worldwide since 1953. With a wide range of surface areas and treatments, we have products for all your basic or advanced cell-based research.
Features:
  • Promote larger plaques used for efficient library screening
  • Compatible with automated systems
  • Sterile; disposable
  • Deep Petri dishes allow for longer culture periods
Ordering Information:
12 per sealed plastic sleeve.
Specifications
Specifications
Sterile