Learn More
Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface.
£462.00 - £885.00
Specifications
Type | Cell Culture Dish |
---|---|
Surface Treatment | UpCell, temperature responsive |
Sterility | Sterile |
Well Shape | Round, Flat Bottom (F-well) |
Product Code | Brand | Description | Culture Area | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Culture Area | Price | Quantity & Availability | ||||
10073833
|
Thermo Scientific™
174902 |
100mm, UpCell dish | 56.7 sq. cm |
£493.00
Pack of 6 |
In Stock
Log in to see stock. |
||||
10592954
|
Thermo Scientific™
174903 |
60mm, UpCell dish | 21.5 sq. cm |
£885.00
Pack of 30 |
In Stock
Log in to see stock. |
||||
10463665
|
Thermo Scientific™
174904 |
35mm, UpCell dish, w/ supportive membrane | 8.8 sq. cm |
£710.00
Pack of 30 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
10165613
|
Thermo Scientific™
174905 |
100mm, UpCell dish w/grid | 56.7 sq. cm |
£462.00
Pack of 6 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
10398963
|
Thermo Scientific™
174906 |
60mm, UpCell dish w/grid | 21.5 sq. cm |
£885.00
Pack of 30 |
Estimated Shipment: 11-02-2025
Log in to see stock. |
||||
Description
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of "cell sheet" with the help of a supportive membrane.
- The hydrophicility of the UpCell surface changes with the external temperature, allowing cells to detach
- No need for dissociation enzymes, ensuring intact cell surface proteins
- No need for physical scraping, resulting in high cell viability
- 3 x 3mm grid design in the 60mm and 100mm dishes prevents cells from clumping and enables harvesting of single cell suspension
- Supportive membranes included in the 35mm dishes allow for harvesting of the cell sheets
- Shelf Life: 3 years from date of manufacturing
Recommended for:
- Culture passaging
- Subsequent single-cell analyses (e.g. flow cytometry)
- 3D tissue modeling for transplantation
- For research use only
Specifications
Cell Culture Dish | |
UpCell, temperature responsive | |
Sterile | |
Round, Flat Bottom (F-well) |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only