Learn More
Thermo Scientific™ Nunc™ Disposable Loops and Needles
BIOPHARMA
Product Code. p-4520792
Click to view available options
Volume:
1 μL
10 μL
Quantity:
Carton of 1000
Carton of 600
Unit Size:
Pack of 1000
Pack of 2400
Pack of 4000
Pack of 600
Product Code. 10048750
Brand: Thermo Scientific™ 251586
Only null left
Add to basket
Edge
Add to basket
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1 μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Volume 10 μL
Material Polystyrene
Sterility Sterile
Color Blue
Product Type Inoculating Loop (Blue)
Quantity Carton of 1000