Learn More
Thermo Scientific™ 96 Well Edge Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10

Color:Clear; Sterility:Non-sterile; Lid:Without Lid
Brand: Thermo Scientific™ 267566
265.66 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge; 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Features- The Edge 2.0 Plate provides an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
- The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL sterile water, buffer or media.
- Replenish fluid in the reservoirs every 3 days in the humidified incubator to ensure the effectiveness of the evaporation barrier.
- Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth.
- The non-treated surface is available for suspension cell culture or general solution-based assays
- Use the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader. The filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.

Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
0.35 cm2/well | |
10 Pk., 160 Cases | |
Clear | |
Low Flange | |
400 μL | |
Non-sterile | |
Flat Bottom (F-well) | |
160 |
Without Lid | |
8 x 12 | |
Nunc | |
Microplate | |
Polystyrene | |
96 | |
Non-treated | |
Less waste, fewer resources used | |
10 |
Videos
-
Thermo Scientific Nunc Edge 2.0 plate
-
Thermo Scientific Nunc Edge 2.0 plate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.