Learn More
Thermo Scientific™ 96 Well Edge Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 1

Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate.
Brand: Thermo Scientific™ 267576
82.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.

Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
0.35 cm2/well | |
1/Pk., 50/Cs. | |
Clear | |
Low Flange | |
400 ÎĽL | |
Sterile | |
Flat Bottom (F-well) | |
50 |
Without Lid | |
8 x 12 | |
Nunc | |
Microplate | |
Polystyrene | |
96 | |
Non-treated | |
Less waste, fewer resources used | |
1 |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Thermo Scientific Nunc Edge 2.0 plate
-
Thermo Scientific Nunc Edge 2.0 plate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.