Learn More
Thermo Scientific™ Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates
Use these well plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays.
Brand: Thermo Scientific™ 437796
155.24 GBP valid until 2024-06-30
Use promo code "21639" to get your promotional price.
Code : 76
Additional Details : Weight : 2.95200kg
Description
Use Thermo Scientific™ Nunc FluoroNunc/LumiNunc 96-Well Solid plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays. Thermo Scientific Nunc Immuno plates offer lot-to-lot reliability, well-to-well reproducibility, and are backed by industry-leading quality standards and certifications.
Features:
- Plate has SBS footprint—fits standard equipment
- Available with flat-bottom or C-shaped wells
- Choice of PolySorp™ or MaxiSorp™ surface
- Offered as black, white or clear plates
- Certified binding homogeneity/reproducibility
Choice of Surface:
PolySorp - Hydrophobic surface preferentially binds biomolecules that have hydrophobic domains.
MaxiSorp - Hydrophilic surface is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character. Available with or without lot specific.
- Performance testing via an IgG binding assay (well-to-well %CV less than 5% for clear plates and less than 10% for colored plates). Tested items come with a certificate.
White:
- Gives maximum reflection
- Minimum autofluorescence and autoluminescence
Black:
- Minimizes background in fluorescence reading
- Minimizes back-scattered light
- Minimizes crosstalk
Clear:
- For time-resolved fluorescence
Compatible with:
Compatible with lids for MicroWell™ plates, sealing tape and breathable membrane
Specifications
Hydrophilic | |
Standard | |
2.5 cm2 | |
8 x 12 | |
Adsorption | |
White | |
Polystyrene | |
250 μL | |
Round, C-bottom | |
5 | |
Antibodies (Glycoproteins) |
Certified for the absorption of IgG; well to well %CV of less than 10%. | |
With Lid | |
Chemiluminescent | |
60 Cases | |
Nunc | |
Microplate | |
96 | |
MaxiSorp | |
60 | |
None |