Learn More
Thermo Scientific™ Nunc™ Immuno Stick
Use the Thermo Scientific™ Nunc™ Immuno Stick in biotechnology, pharmaceutical laboratories or in production of vaccines and diagnostic kits.
Brand: Thermo Scientific™ 472230
925.00 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : A0
Additional Details : Weight : 5.36000kg
Description
Use the Thermo Scientific™ Nunc™ Immuno Stick in biotechnology, pharmaceutical laboratories or in production of vaccines and diagnostic kits. The system consists of a tube and immuno stick with a paddle.
Features:
- Choice of Thermo Scientific™ MaxiSorp™ or PolySorb™
- Polypropylene tube for minimal adsorption of assay components
- Polystyrene paddle, surface area, 5.2cm2
- Visual result: depostited substrate or colored solution
- External dimensions (L×W): 1.93 × 0.47 in. (49 × 12mm)
Ordering Information:
Application literature is available.
Note:
The manufacture and use of synthetic materials for solid phase immuno assays as covered under EU Patent No. 0126392 and US Patent No. 4,980,299 is licensed to Nunc A/S. Materials are manufactured under licence from Ventrex Laboratories, Inc., US Patent No. 4,225,575.
Specifications
MaxiSorp paddle surface | |
Immuno Stick | |
1800/Cs. |