Learn More
Thermo Scientific™ Nunc™ IVF Dishes

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.
Brand: Thermo Scientific™ 150255
239.62 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.
Product features for reliable results:
- Facilitate gas exchange with vented lid
- Available in 35mm, 60mm and 90mm sizes
- Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF:
- CE-marked (Notified Body 2460), FDA-listed
- Produced under ISO 13485 with full lot traceability
- Sterile (SAL 10-6)
- Non-pyrogenic (LAL tested)
- USP Class VI polystyrene
- Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Warranty: 90 days

Specifications
CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested | |
8.8 cm2 | |
Sterile | |
500 | |
11 mm |
IVF Dish, 35mm, Lid w/Airvent | |
Polystyrene (USP Class VI) | |
Non-treated | |
10 | |
35 mm |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
184891 | Certificate of Analysis | 14/02/2025 | 150255 |
184767 | Certificate of Analysis | 11/01/2025 | 150255 |
184891 | Certificate of Irradiation | 20/01/2025 | 150255 |
184767 | Certificate of Irradiation | 08/01/2025 | 150255 |
184766 | Certificate of Irradiation | 06/01/2025 | 150255 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.