Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide System
Descripción
Grow, stain, and examine cells directly on the Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide™, which consists of a polystyrene media chamber attached to a microscopic slide. Slide surface is specially treated for adherent cell culture. Choose either specialty washed Soda Lime Glass slide or cell attachment optimized Permanox Plastic slide. Media chamber is easily removable without the need for a tool.
- Broad range of formats and well number to meet application needs
- Available with a Permanox solvent-resistant plastic slide for adherent cell culture
- Writing surface provides easy identification of samples
- Save time and reagents by growing cells directly on microscope slide
- In 1, 2, 4, 8 and 16 well configurations; all measure 25 × 75mm
- Medical-grade silicone sealing gasket can be removed for cover slipping
- No cell transfer needed prior to visualization/staining
- Useful for viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology
- Suitable for use with fluorescent labels
- Sterile
Especificaciones
Especificaciones
| Wells | 1 |
| Culture Area | 9.4 cm2 |
| Lid Material | Polystyrene |
| Description | 1-well Chamber Slide w/ removable well |
| Surface Treatment | Nunclon Delta |
| Material | Permanox Plastic |
| Volume (Metric) Working | 2.5 to 4.5 mL |
| Includes | Clear polystyrene lid with handle on the side for aseptic manipulations |
| No. per Pack | 16 |
| Dimensions (LxW) | 75 x 25 mm |
| Mostrar más |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.