missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide System

Product Code. 11307774
Click to view available options
Wells:
1
2
4
8
16
Material:
Permanox Plastic
Soda Lime Glass
Unit Size:
Pack of 16
Pack of 96
This item is not returnable. View return policy

Product Code. 11307774

Brand: Thermo Scientific™ 177445

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Grow anchorage dependent cells right on the microscope slide using the unique removable chamber of the Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide System.

Grow, stain, and examine cells directly on the Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide™, which consists of a polystyrene media chamber attached to a microscopic slide. Slide surface is specially treated for adherent cell culture. Choose either specialty washed Soda Lime Glass slide or cell attachment optimized Permanox Plastic slide. Media chamber is easily removable without the need for a tool.

  • Broad range of formats and well number to meet application needs
  • Available with a Permanox solvent-resistant plastic slide for adherent cell culture
  • Writing surface provides easy identification of samples
  • Save time and reagents by growing cells directly on microscope slide
  • In 1, 2, 4, 8 and 16 well configurations; all measure 25 × 75mm
  • Medical-grade silicone sealing gasket can be removed for cover slipping
  • No cell transfer needed prior to visualization/staining
  • Useful for viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology
  • Suitable for use with fluorescent labels
  • Sterile
TRUSTED_SUSTAINABILITY

Specifications

Wells 8
Culture Area 0.8 cm2
Lid Material Polystyrene
Description 8-well Chamber Slide w/ removable wells
Surface Treatment Nunclon Delta
Material Permanox Plastic
Volume (Metric) Working 0.2 to 0.4 mL
Includes Clear polystyrene lid with handle on the side for aseptic manipulations
No. per Pack 16
Dimensions (LxW) 75 x 25 mm
Sterility Sterile
Closure Color Clear
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.