Learn More
Thermo Scientific™ Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Brand: Thermo Scientific™ 253623
272.17 GBP valid until 2025-05-31
Use promo code "25075" to get your promotional price.
Description
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
- Lids are for use with any standard plates
- 96 Well Lids have individual condensation rings
- Lids have notches for quick orientation
- Universal Lids can be used on any standard height plate on the market

Specifications
Clear | |
Polystyrene | |
384/1536 | |
5 in. | |
Yes | |
100 | |
5 x 3.35 in. (127 x 85 mm) | |
3.35 in. | |
LowBot Lids | |
None | |
All well shapes | |
No |
5 x 3.35 x 0.18 in. (127 x 85 x 4.6 mm) | |
Non-sterile | |
4.6 mm | |
127 mm | |
0.18 in. | |
25 | |
Yes | |
85 mm | |
Nunc | |
-20°C to +80°C | |
All Microplates |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1292030 | Certificate of Compliance | 11/12/2024 | 253623 |
1376715 | Certificate of Compliance | 11/12/2024 | 253623 |
1299945 | Certificate of Compliance | 04/11/2020 | 253623 |
1283106 | Certificate of Compliance | 06/05/2020 | 253623 |
1281737 | Certificate of Compliance | 09/04/2020 | 253623 |
Special Offers
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-03-31
-
Multiple Manufacturers
New Offer Shop Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.