Learn More
Thermo Scientific™ Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Brand: Thermo Scientific™ 253624
284.57 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
- Lids are for use with any standard plates
- 96 Well Lids have individual condensation rings
- Lids have notches for quick orientation
- Universal Lids can be used on any standard height plate on the market

Specifications
Clear | |
Polystyrene | |
384/1536 | |
5 in. | |
Yes | |
100 | |
5 x 3.35 in. (127 x 85 mm) | |
3.35 in. | |
LowBot Lids | |
None | |
All well shapes | |
No |
5 x 3.35 x 0.18 in. (127 x 85 x 4.6 mm) | |
Sterile | |
4.6 mm | |
127 mm | |
0.18 in. | |
25 | |
Yes | |
85 mm | |
Nunc | |
-20°C to +80°C | |
All Microplates |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1398657 | Certificate of Processing | 04/12/2024 | 253624 |
1299455 | Certificate of Processing | 04/12/2024 | 253624 |
1272427 | Certificate of Processing | 04/12/2024 | 253624 |
1381255 | Certificate of Processing | 04/12/2024 | 253624 |
1273586 | Certificate of Processing | 04/12/2024 | 253624 |
Special Offers
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30
Days
91Hours
14Minutes
1Seconds
20 -
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
13Hours
14Minutes
1Seconds
20New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.