Learn More
Thermo Scientific™ Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Brand: Thermo Scientific™ 250002
122.21 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
- Lids are for use with any standard plates
- 96 Well Lids have individual condensation rings
- Lids have notches for quick orientation
- Universal Lids can be used on any standard height plate on the market

Specifications
Clear | |
Polystyrene | |
384/1536 | |
5 in. | |
No | |
60 | |
5 x 3.35 in. (127 x 85 mm) | |
3.35 in. | |
Universal Lid | |
None | |
All well shapes | |
No |
5 x 3.35 x 0.33 in. (127 x 85 x 8.34 mm) | |
Sterile | |
8.34 mm | |
127 mm | |
0.33 in. | |
20 | |
No | |
85 mm | |
Nunc | |
-20°C to +80°C | |
All Microplates |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.