missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Microplate Lids
GREENER_CHOICE

Product Code. 10393072
Click to view available options
Plate Compatibility:
384-well Plate
384/1536-well Plate, OmniTray
96-well Plate
All Microplates
Sterility:
Non-sterile
Sterile
Condensation Rings:
No
Yes
Cut-off Corners:
Long Side
No
Unit Size:
Pack of 100
Pack of 120
Pack of 180
Pack of 50
Pack of 60
This item is not returnable. View return policy

Product Code. 10393072

Brand: Thermo Scientific™ 264612

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.

Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.

  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Dimensions (L x W x H) 5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
Material Polystyrene
Sterility Non-sterile
No. of Wells 384
Height (Metric) 7.9 mm
Length (English) 5 in.
Length (Metric) 127 mm
Condensation Rings No
Height (English) 0.3 in.
No. per Case 180
No. per Pack 20
Dimensions (L x W) 5 x 3.35 in. (127 x 85 mm)
Evaporation Barrier Yes
Width (English) 3.35 in.
Width (Metric) 85 mm
Product Type Plate Lid
Product Line Nunc
Adhesive None
Temperature Range (Metric) -20°C to +80°C
Well Shape All well shapes
Plate Compatibility 384-well Plate
Cut-off Corners Long Side
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.