Learn More
Thermo Scientific™ Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Brand: Thermo Scientific™ 448143
Description
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
- Lids are for use with any standard plates
- 96 Well Lids have individual condensation rings
- Lids have notches for quick orientation
- Universal Lids can be used on any standard height plate on the market

Specifications
White | |
Polystyrene | |
384/1536 | |
5 in. | |
No | |
120 | |
No | |
85 mm | |
Plate Cover | |
Long Side |
5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm) | |
Non-sterile | |
7.9 mm | |
127 mm | |
0.3 in. | |
20 | |
3.35 in. | |
Low Profile | |
384/1536-well Plate, OmniTray |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1321718 | Certificate of Compliance | 09/08/2021 | 448143 |
1314133 | Certificate of Compliance | 07/04/2021 | 448143 |
1292728 | Certificate of Compliance | 15/09/2020 | 448143 |
1278963 | Certificate of Compliance | 11/03/2020 | 448143 |
1227818 | Certificate of Compliance | 11/04/2018 | 448143 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.