Learn More
Thermo Scientific™ Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Brand: Thermo Scientific™ 264616
224.94 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
- Lids are for use with any standard plates
- 96 Well Lids have individual condensation rings
- Lids have notches for quick orientation
- Universal Lids can be used on any standard height plate on the market

Specifications
Clear | |
Polystyrene | |
384 | |
5 in. | |
No | |
180 | |
5 x 3.35 in. (127 x 85 mm) | |
3.35 in. | |
Plate Lid | |
None | |
All well shapes | |
Long Side |
5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm) | |
Sterile | |
7.9 mm | |
127 mm | |
0.3 in. | |
20 | |
Yes | |
85 mm | |
Nunc | |
-20°C to +80°C | |
384-well Plate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1375986 | Certificate of Compliance | 11/12/2024 | 264616 |
1335414 | Certificate of Compliance | 11/12/2024 | 264616 |
1297559 | Certificate of Processing | 04/12/2024 | 264616 |
1281773 | Certificate of Processing | 04/12/2024 | 264616 |
1295866 | Certificate of Processing | 04/12/2024 | 264616 |
Special Offers
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30
Days
91Hours
13Minutes
58Seconds
29
Your input is important to us. Please complete this form to provide feedback related to the content on this product.