missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Microplate Plastic Storage Racks

Conveniently and safely store and transport samples in microplate, deep-well plates and cryobank racks by using Thermo Scientific™ Nunc™ Plastic Storage Racks.

Brand:  Thermo Scientific™ 344380

 View more versions of this product

Product Code. 12736558

  • £445.00 / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Conveniently and safely store and transport samples in microplate, deep-well plates and cryobank racks by using Thermo Scientific™ Nunc™ Plastic Storage Racks, ideal for genome library storage, DNA and RNA libraries, and storage of compounds for high throughput screening.

  • Horizontal design fits chest, upright freezers and incubator
  • Retainer clip holds plates securely in place
  • Handles on top and side for easy transport
  • Locking bar keeps plates secure
  • Handle allows convenient transport
  • Freezer compatible down to -80°C
  • Not for use in liquid/vapor nitrogen
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Microplate Rack
Green
Polypropylene, Metal
51 cm
2 in.
14 cm
5 x 5
Cryobank Racks, Deepwell Microplates, Microplates
Rectangular
25 cm
5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.