Learn More
Thermo Scientific™ 96 Well White Plate, Coverglass Bottom, TC Surface, Pack of 6
Color:White, Clear Bottom; Sterility:Sterile; Lid:With Lid; Type: Optical Coverglass Base
Brand: Thermo Scientific™ 164590
2656.79 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
- Background, autoflorescence and crosstalk are improved
- Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
- White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
- Cell culture surface is ideal for cell-based assays
- CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
- Customized bar coding available

Specifications
Custom Barcoding Available | |
With Lid | |
6/Pk., 30/Cs. | |
White | |
High Flange | |
400 μL | |
Sterile | |
Flat Bottom (F-well) | |
6 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
8 x 12 | |
Nunc | |
Optical Coverglass Base | |
Polystyrene | |
96 | |
Cell Culture | |
30 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1323688 | Certificate of Compliance | 11/12/2024 | 164590 |
1364525 | Certificate of Compliance | 17/09/2022 | 164590 |
1368557 | Certificate of Compliance | 15/09/2022 | 164590 |
1331709 | Certificate of Compliance | 24/02/2022 | 164590 |
1315271 | Certificate of Compliance | 01/04/2021 | 164590 |
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
More Discovery, Less Cost
Save on thousands of eligible products.Promo Code: 24111Expires: 2025-03-21
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.