Learn More
Thermo Scientific™ 96 Well Plate, Non-Treated Surface, No lid, Non-sterile, Pack of 5
These microplates are optimized for robotics, automated readers and liquid handling systems. Ideal for antibiotic screens, cell-based assays and screening compounds.
Brand: Thermo Scientific™ 269620
78.60 GBP valid until 2024-12-31
Use promo code "21894" to get your promotional price.
Code : 76
Additional Details : Weight : 2.86110kg
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Custom Barcoding Available | |
Without Lid | |
8 x 12 | |
Nunc | |
Microplate | |
96 | |
Non-treated | |
60 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
96F well plate, Non-Treated, clear, without lid, Non-sterile (Pack of 5, Case of 60) | |
5/Pk., 60/Cs. | |
Clear | |
Polystyrene | |
Non-sterile | |
Flat Bottom (F-well) | |
5 |