Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Non-Treated, Flat-Bottom Microplate

Color:Clear; Sterility:Sterile; Lid:Without Lid
Brand: Thermo Scientific™ 256510
194.83 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available

Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Without Lid | |
8 x 12 | |
Nunc | |
Microplate | |
96 | |
Non-treated | |
180 |
Standard | |
F 96 well plate, Non-Treated, clear, without lid, Sterile (Pack of 10, Case of 180) | |
10 Pk., 180 Cases | |
Clear | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
10 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
179031 | Certificate of Compliance | 03/12/2024 | 256510 |
166656 | Certificate of Compliance | 03/12/2024 | 256510 |
173572 | Certificate of Compliance | 03/12/2024 | 256510 |
123456 | Certificate of Compliance | 17/10/2024 | 256510 |
179031 | Certificate of Compliance | 16/10/2024 | 256510 |
Special Offers
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.