missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 96 Well Plate, Non-Treated, Non-Sterile, Pack of 10
Color:Clear; Sterility:Non-sterile; Lid:With Lid
Brand: Thermo Scientific™ 260895
222.88 GBP valid until 2024-10-31
Use promo code "21744" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Custom Barcoding Available | |
With Lid | |
8 x 12 | |
Nunc | |
Microplate | |
400 μL | |
Non-sterile | |
Flat Bottom (F-well) | |
10 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
96F well plate, Non-Treated, clear, with lid, Non-sterile (Pack of 10, Case of 160) | |
10 Pk., 160 Case | |
Clear | |
Polystyrene | |
96 | |
Non-treated | |
160 | |
160/Cs. |