missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom, Optical Coverglass Base Microplate

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid; Type: Optical Coverglass Base

Brand:  Thermo Scientific™ 164588

1943.43 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.


Product Code. 10472222

  • £2915.00 / Pack of 30
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.

Features:

  • Background, autoflorescence and crosstalk are improved
  • Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
  • White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
  • Cell culture surface is ideal for cell-based assays
  • CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
  • Customized bar coding available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
8 x 12
Nunc
Microplate
96
Cell Culture
30
Case of 30
With Lid
6 Pk., 30 Cases
Black, Clear Bottom
Polystyrene
Sterile
Flat Bottom (F-well)
6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1419511 Certificate TypeCertificate of Compliance Date14/02/2025 Catalog Number164588
Lot Number1407550 Certificate TypeCertificate of Compliance Date11/02/2025 Catalog Number164588
Lot Number1411700 Certificate TypeCertificate of Compliance Date05/12/2024 Catalog Number164588
Lot Number1411699 Certificate TypeCertificate of Compliance Date27/11/2024 Catalog Number164588
Lot Number1406078 Certificate TypeCertificate of Compliance Date30/05/2024 Catalog Number164588
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom, Optical Coverglass Base Microplate > Coverglass base; with Lid; Black; Cell culture surface

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.