Learn More
Thermo Scientific™ 96 Black Plate, Non-Treated Coverglass Surface, No Lid, Non-Sterile, Pack of 5
Product Code. 10194202
Click to view available options
Quantity:
Case of 30
Unit Size:
Pack of 30
Product Code. 10194202
Brand: Thermo Scientific™ 265300
Only null left
Add to basket
Add to basket
Color:Black, Clear Bottom; Sterility; Non-sterile; Lid, Without Lid; Type: Optical Coverglass Base
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
  • Background, autoflorescence and crosstalk are improved
  • Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
  • White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
  • Cell culture surface is ideal for cell-based assays
  • CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
  • Customized bar coding available
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Lid Without Lid
Array 8 x 12
Packaging 5/Pk., 30/Cs.
Product Line Nunc
Color Black
Product Type Optical Coverglass Base
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 30
No. per Pack 5
Quantity Case of 30