missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ MicroWell™ MiniTrays
Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
£346.00 - £1610.00
Specifications
Bottom Shape | Flat |
---|---|
Material | Polystyrene |
Shape | Rectangular |
Type | Cell Culture Dish |
Length (Metric) | 84 mm |
Product Code | Brand | Description | Well Count | Closure Included | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Well Count | Closure Included | Price | Quantity & Availability | |||
10192831
|
Thermo Scientific™
438733 |
MicroWell MiniTray for serological applications | 72 | Closure Included |
£346.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10104371
|
Thermo Scientific™
439225 |
MicroWell MiniTray for serological applications | 60 | Closure Included |
£346.00
Pack of 100 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||
10204221
|
Thermo Scientific™
452256 |
MicroWell MiniTray | 60 | Closure Included |
£1490.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
10112791
|
Thermo Scientific™
470378 |
MicroWell MiniTray | 72 | Closure Included |
£1610.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
10326141
|
Thermo Scientific™
472400 |
MicroWell MiniTray | 60 | Closure Not Included |
£805.00
Pack of 400 |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||
Description
Create more space in lab freezers and inubators with Thermo Scientific™ Nunc™ MicroWell™ MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
- Limited adsorption of proteins
- Lids have keyed tabs for proper orientation
- Large raised alphanumeric coordinates
- Unique surface treatment to maximize sample and reagent mixing
- MiniTrays for serological applications have slightly smaller well dimensions
Specifications
Flat | |
Rectangular | |
84 mm | |
59 mm |
Polystyrene | |
Cell Culture Dish | |
59 mm |