Learn More
Thermo Scientific™ Nunc™ Multidishes with UpCell™ Surface

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins.
£322.00 - £344.00
Specifications
Type | Multi Dish |
---|---|
No. per Pack | 1 |
No. per Case | 6 |
Color | Clear |
Sterility | Sterile |
Product Code | Brand | No. of Wells | Culture Area | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Culture Area | Price | Quantity & Availability | |||||
10616234
![]() |
Thermo Scientific™
174898 |
48 | 1.1 sq. cm |
£331.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
10532204
![]() |
Thermo Scientific™
174899 |
24 | 1.9 sq. cm |
£322.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
10288143
|
Thermo Scientific™
174900 |
12 | 3.5 sq. cm |
£344.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
10542204
![]() |
Thermo Scientific™
174901 |
6 | 9.6 sq. cm |
£340.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
Description
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of cell sheet with the help of a supportive membrane.
Features:- The hydrophicility of the UpCell surface changes with the external temperature, allowing cells to detach
- No need for dissociation enzymes, ensuring intact cell surface proteins
- No need for physical scraping, resulting in high cell viability
- Supportive membranes included in the 6-well plates allow for harvesting of the cell sheets
- Customized bar coding available
- Shelf Life: 3 years from date of manufacturing
Recommended for:
- Culture passaging
- Subsequent single-cell analyses (e.g. flow cytometry)
- 3D tissue modeling for transplantation
- For research use only
Specifications
Multi Dish | |
6 | |
Sterile |
1 | |
Clear | |
Rectangular, Flat Bottom (F-well) |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.