Learn More
Thermo Scientific™ Nunc™ PETG Roller Bottles
Description
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.
- Molded of durable PETG
- Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
- Provides solutions for all scales with wide range of surface areas from 1050cm2 to 4200cm2
- Reduces wrist strain and increases productivity with quick-action ergonomic closure
- Can be traced by lot number on every bottle
- Cell culture expansion
- Vaccine production
- Antibody production

Specifications
Specifications
Bottle Surface | Pleated |
Color | Clear |
Description | 2160-20 |
Material | PETG |
No. per Case | 20 |
No. per Pack | 20 |
Packaging | 20/case |
Cell Growth Area | 2100 cm2 |
Closure Included | Yes |
Graduated | Yes |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Documents
Scientific Resources
Application Notes
Brochures
Product Information
Manuals
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1421599 | Certificate of Processing | 03/02/2025 | 2160-20 |
1420301 | Certificate of Processing | 30/01/2025 | 2160-20 |
1420300 | Certificate of Processing | 23/01/2025 | 2160-20 |
1420299 | Certificate of Processing | 20/01/2025 | 2160-20 |
1421599 | Certificate of Compliance | 18/01/2025 | 2160-20 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.