missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ PETG Roller Bottles
144.88 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Cell Growth Area:
1050 cm2
1700 cm2
1800 cm2
2100 cm2
4200 cm2
Bottle Surface:
Pleated
Smooth
Packaging:
20/case
22/case
5/sleeve, 20/case
Description
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.
Key Features:
- Molded of durable PETG
- Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
- Provides solutions for all scales with wide range of surface areas from 1050cm2 to 4200cm2
- Reduces wrist strain and increases productivity with quick-action ergonomic closure
- Can be traced by lot number on every bottle
- Cell culture expansion
- Vaccine production
- Antibody production
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Bottle Surface | Smooth |
Color | Clear |
Description | 1060-20 |
Material | PETG |
No. per Case | 20 |
No. per Pack | 20 |
Packaging | 20/case |
Cell Growth Area | 1050 cm2 |
Closure Included | Yes |
Graduated | Yes |
Show More |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'