Learn More
Thermo Scientific™ Nunc™ Polycarbonate Cell Culture Inserts in Multi-Well Plates
BIOPHARMA
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Brand:  Thermo Scientific™ 140654
 View more versions of this product
Product Code. 10504394
  • £400.00 / Pack of 48
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Features:
  • Thermo Scientific™ Nunclon™ Delta-treated and tested for cell attachment
  • Tabs provide better grip and easy handling
  • Nontoxic and nonpyrogenic
Ordering Information:
Packed in resealable bags
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polycarbonate Cell Culture Inserts with 3 micron pore size in 12-well plate
1.13 cm2
1.1 mL
3 μm
Multidish 12
48
Clear
Sterile
12
Polycarbonate (Membrane)
<1.7 x 106 pores/cm2
Culture Insert
12
12-Well Plate
Cell culture-treated
12
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.