Learn More
Thermo Scientific™ Nunc™ Polycarbonate Cell Culture Inserts in Multi-Well Plates
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Brand:  Thermo Scientific™ 140644
 View more versions of this product
Product Code. 10637143
  • £234.00 / Pack of 24
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Features:
  • Thermo Scientific™ Nunclon™ Delta-treated and tested for cell attachment
  • Tabs provide better grip and easy handling
  • Nontoxic and nonpyrogenic
Ordering Information:
Packed in resealable bags
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polycarbonate Cell Culture Inserts with 8 micron pore size in 6-well plate
3.14 cm2
1.5 mL
8 μm
Multidish 6
24
Clear
Sterile
6
Polycarbonate (Membrane)
<0.85 x 105 pores/cm2
Culture Insert
6
6-Well Plate
Cell culture-treated
6
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.