Learn More
Thermo Scientific™ Nunc™ Polystyrene Roller Bottles
Description
Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6
- One-piece seamless design: Reduces possibility of leaking through a seam
- Shallow and deep indents: Unique ergonomic design makes the roller bottle excellent for both manual and automated handling
- Proprietary manufacturing process: Greater impact resistance
- Vertical expanded surfaces version: Vertical pleat orientation facilitates emptying and reduces retention of product
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Bottle Surface | Smooth surface, deep indent |
Closure Material | HDPE |
Closure Type | EZ Twist Vented Cap |
Product Type | Roller Bottle |
Color | Clear |
For Use With (Application) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 2 |
Packaging | 2/Pk., 20/Case |
Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.